Assay Details
| Gene Symbol: RPP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244694.1 | NP_001231623.1 | 4 - 5 | 446 | 101 | |
| XM_005653456.2 | XP_005653513.1 | 422 | 101 | |||
| GenBank mRNA | AK399671.1 | - | 4 - 5 | 441 | 101 | |
| EW455534.2 | - | 4 - 5 | 395 | 101 | ||
More Information
Set Membership: |
|

RPP21
Genomic Map