Assay Details
| Gene Symbol: WRAP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244721.1 | NP_001231650.1 | 8 - 9 | 864 | 58 | |
| XM_005664956.3 | XP_005665013.1 | 807 | 58 | |||
| XM_005664957.3 | XP_005665014.1 | 894 | 58 | |||
| GenBank mRNA | AK347555.1 | - | 8 - 9 | 864 | 58 | |
More Information
Set Membership: |
|

WRAP73
Genomic Map