Assay Details
| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244730.1 | NP_001231659.1 | 6 - 7 | 881 | 82 | |
| XM_005653697.3 | XP_005653754.1 | 946 | 82 | |||
| XM_005653698.3 | XP_005653755.1 | 3108 | 82 | |||
| XM_005653699.3 | XP_005653756.1 | 937 | 82 | |||
| XM_005653701.3 | XP_005653758.1 | 976 | 82 | |||
| XM_013977422.2 | XP_013832876.1 | 1420 | 82 | |||
| XM_013977439.2 | XP_013832893.1 | 1016 | 82 | |||
| XM_013977441.1 | XP_013832895.1 | 1026 | 82 | |||
| XM_021102035.1 | XP_020957694.1 | 1483 | 82 | |||
| XM_021102036.1 | XP_020957695.1 | 1678 | 82 | |||
| GenBank mRNA | AK238962.1 | - | 6 - 7 | 881 | 82 | |
| AK398411.1 | - | 4 - 5 | 1259 | 82 | ||
More Information
Set Membership: |
|

JAML
Genomic Map