Assay Details
| Gene Symbol: JAML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244730.1 | NP_001231659.1 | 9 - 10 | 1174 | 92 | |
| XM_005653697.3 | XP_005653754.1 | 1239 | 92 | |||
| XM_005653698.3 | XP_005653755.1 | 3401 | 92 | |||
| XM_005653699.3 | XP_005653756.1 | 1230 | 92 | |||
| XM_005653701.3 | XP_005653758.1 | 1269 | 92 | |||
| XM_013977422.2 | XP_013832876.1 | 1713 | 92 | |||
| XM_013977439.2 | XP_013832893.1 | 1309 | 92 | |||
| XM_013977441.1 | XP_013832895.1 | 1319 | 92 | |||
| XM_021102035.1 | XP_020957694.1 | 1776 | 92 | |||
| XM_021102036.1 | XP_020957695.1 | 1971 | 92 | |||
| GenBank mRNA | AK238962.1 | - | 9 - 10 | 1174 | 92 | |
| AK389867.1 | - | 9 - 10 | 1145 | 92 | ||
| AK398411.1 | - | 7 - 8 | 1552 | 92 | ||
More Information
Set Membership: |
|

JAML
Genomic Map