Assay Details
| Gene Symbol: INMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244733.1 | NP_001231662.1 | 2 - 3 | 418 | 65 | |
| GenBank mRNA | AK232648.1 | - | 2 - 3 | 391 | 65 | |
| AK350501.1 | - | 2 - 3 | 418 | 65 | ||
More Information
Set Membership: |
|

INMT
Genomic Map