Assay Details
| Gene Symbol: MTFMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244826.1 | NP_001231755.1 | 5 - 6 | 763 | 69 | |
| XM_005659485.3 | XP_005659542.1 | 781 | 69 | |||
| XM_013992885.2 | XP_013848339.1 | 781 | 69 | |||
| GenBank mRNA | AK232162.1 | - | 5 - 6 | 760 | 69 | |
| AK233774.1 | - | 5 - 6 | 765 | 69 | ||
More Information
Set Membership: |
|

MTFMT
Genomic Map