Assay Details
| Gene Symbol: KRT31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244895.1 | NP_001231824.1 | 6 - 7 | 1157 | 107 | |
| GenBank mRNA | AK349827.1 | - | 6 - 7 | 1159 | 107 | |
| AK349901.1 | - | 5 - 6 | 631 | 107 | ||
| AK397410.1 | - | 4 - 5 | 587 | 107 | ||
| AK400905.1 | - | 1157 | 107 | |||
More Information
Set Membership: |
|

KRT31
Genomic Map