Assay Details
| Gene Symbol: CLDN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160075.1 | NP_001153547.1 | 1 - 2 | 486 | 122 | |
| GenBank mRNA | FJ887981.1 | - | 1 - 2 | 486 | 122 | |
More Information
Set Membership: |
|

CLDN3
Genomic Map