Assay Details
| Gene Symbol: UBD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160088.1 | NP_001153560.1 | 2 | 321 | 129 | |
| GenBank mRNA | AK231383.1 | - | 2 | 345 | 129 | |
| AK345111.1 | - | 2 | 400 | 129 | ||
More Information
Set Membership: |
|

UBD
Genomic Map