Assay Details
| Gene Symbol: POLR1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243956.1 | NP_001230885.1 | 4 | 2517 | 171 | |
| GenBank mRNA | AK238629.1 | - | 1 | 1072 | 171 | |
More Information
Set Membership: |
|

POLR1F
Genomic Map