Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244524.1 | NP_001231453.1 | 10 | 1619 | 83 | |
| XM_005666793.2 | XP_005666850.1 | 2204 | 83 | |||
| GenBank mRNA | AY609550.1 | - | 10 | 1544 | 83 | |
| BM659162.1 | - | 1 | 164 | 83 | ||
More Information
Set Membership: |
|

FGG
Genomic Map