Assay Details
| Gene Symbol: NRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656275.3 | XP_005656332.1 | 654 | 87 | ||
| GenBank mRNA | AK346404.1 | - | 7 - 8 | 714 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656275.3 | XP_005656332.1 | 654 | 87 | ||
| GenBank mRNA | AK346404.1 | - | 7 - 8 | 714 | 87 | |
Set Membership: |
|