Assay Details
| Gene Symbol: MIR148B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035387.1 | 1 | 33 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
nuclear factor, erythroid 2 |
NFE2 | -29.2kb | Anti-sense | Chr. 5 - 19494672 - 19501810 on NCBI Build Sscrofa11.1 | |
| Downstream | G protein-coupled receptor 84 | GPR84 | 21.8kb | Anti-sense | Chr. 5 - 19552828 - 19554848 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR148B
Genomic Map