Assay Details
Gene Symbol: MIR1224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049179.1 | 1 | 53 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATP binding cassette subfamily F member 3 | ABCF3 | -28.1kb | Sense | Chr. 13 - 122102939 - 122112896 on NCBI Build Sscrofa11.1 | |
Downstream | ALG3 alpha-1,3- mannosyltransferase | ALG3 | 895.0 | Anti-sense | Chr. 13 - 122142044 - 122148777 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
