Assay Details
| Gene Symbol: MIR383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049181.1 | 1 | 48 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DLC1 Rho GTPase activating protein | DLC1 | -1.1mb | Sense | Chr. 17 - 1169709 - 1637171 on NCBI Build Sscrofa11.1 | |
| Downstream | tumor suppressor candidate 3 | TUSC3 | 614.6kb | Anti-sense | Chr. 17 - 3312684 - 3507372 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR383
Genomic Map