Assay Details
| Gene Symbol: MIR1249-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128444.1 | 1 | 40 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KIAA0930 | KIAA0930 | Sense | Chr. 5 - 4152371 - 4198372 on NCBI Build Sscrofa11.1 | ||
| Downstream | nucleoporin 50 | NUP50 | 6.3kb | Anti-sense | Chr. 5 - 4194308 - 4219410 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR1249-1
Genomic Map