Assay Details
| Gene Symbol: MIRLET7G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038522.1 | 1 | 44 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein phosphatase, Mg2+/Mn2+ dependent 1M |
PPM1M | -20.5kb | Anti-sense | Chr. 13 - 34380518 - 34385600 on NCBI Build Sscrofa11.1 | |
| Downstream | glycerate kinase | GLYCTK | 27.2kb | Anti-sense | Chr. 13 - 34433403 - 34440037 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIRLET7G
Genomic Map