Assay Details
| Gene Symbol: MIR378-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038523.1 | 1 | 44 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho guanine nucleotide exchange factor 37 | ARHGEF37 | -74.2kb | Sense | Chr. 2 - 150662470 - 150751648 on NCBI Build Sscrofa11.1 | |
| Downstream | phosphodiesterase 6A | PDE6A | 100.7kb | Anti-sense | Chr. 2 - 150926652 - 150986050 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR378-1
Genomic Map