Assay Details
Gene Symbol: MIR152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038532.1 | 1 | 40 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CDK5 regulatory subunit associated protein 3 | CDK5RAP3 | -57.8kb | Anti-sense | Chr. 12 - 24224262 - 24234468 on NCBI Build Sscrofa11.1 | |
Downstream |
nuclear factor, erythroid 2 like 1 |
NFE2L1 | 11.1kb | Anti-sense | Chr. 12 - 24303381 - 24316765 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
