Assay Details
Gene Symbol: MIR374B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038552.1 | 1 | 37 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-421 | MIR421 | ssc-mir-421 | -77.0 | Sense | Chr. X - 59550992 - 59551069 on NCBI Build Sscrofa11.1 |
Downstream | microRNA mir-545 | MIR545 | ssc-mir-545 | 51.5kb | Sense | Chr. X - 59602710 - 59602788 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
