Assay Details
Gene Symbol: MIR195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038559.1 | 1 | 40 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102161115 | LOC102161115 | Sense | Chr. 12 - 52420175 - 52424455 on NCBI Build Sscrofa11.1 | ||
Downstream | microRNA mir-497 | MIR497 | ssc-mir-497 | 211.0 | Sense | Chr. 12 - 52422329 - 52422408 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
