Assay Details
| Gene Symbol: MIR1839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_035401.1 | 1 | 37 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibronectin type III and SPRY domain containing 2 | FSD2 | -2.6kb | Anti-sense | Chr. 7 - 52144469 - 52194142 on NCBI Build Sscrofa11.1 | |
| Downstream | adaptor related protein complex 3 subunit beta 2 | AP3B2 | 5.8kb | Anti-sense | Chr. 7 - 52202649 - 52294154 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR1839
Genomic Map