Assay Details
| Gene Symbol: MIR9841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128528.1 | 1 | 43 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuritin 1 | NRN1 | -144.1kb | Sense | Chr. 7 - 3682568 - 3691328 on NCBI Build Sscrofa11.1 | |
| Downstream | lymphocyte antigen 86 | LY86 | MD-1 | 272.6kb | Anti-sense | Chr. 7 - 4108124 - 4156629 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

MIR9841
Genomic Map