Assay Details
Gene Symbol: MIR9860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128556.1 | 1 | 37 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | interferon regulatory factor 2 binding protein like | IRF2BPL | -2.9kb | Sense | Chr. 7 - 100119282 - 100123820 on NCBI Build Sscrofa11.1 | |
Downstream | CLOCK interacting pacemaker | CIPC | KIAA1737 | 53.6kb | Anti-sense | Chr. 7 - 100180389 - 100199529 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
