Assay Details
| Gene Symbol: CYP2C91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132429.1 | 6 - 7 | 978 | 72 | ||
| GenBank mRNA | AK351408.1 | - | 6 - 7 | 859 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATM interactor | LOC102166602 | -37.5kb | Sense | Chr. 14 - 106656553 - 106659220 on NCBI Build Sscrofa11.1 | |
| Downstream | PDZ and LIM domain 1 | PDLIM1 | 45.4kb | Sense | Chr. 14 - 106744320 - 106793606 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

CYP2C91
Genomic Map