Assay Details
Gene Symbol: MIR129A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038576.1 | 1 | 40 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hydroxysteroid 17-beta dehydrogenase 12 | HSD17B12 | -112.5kb | Sense | Chr. 2 - 18461487 - 18611761 on NCBI Build Sscrofa11.1 | |
Downstream | tetratricopeptide repeat domain 17 | TTC17 | 83.2kb | Sense | Chr. 2 - 18807521 - 18954667 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
