Assay Details
Gene Symbol: MIR149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038595.1 | 1 | 42 | 55 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102160973 | LOC102160973 | -3.5kb | Anti-sense | Chr. 15 - 139463791 - 139471262 on NCBI Build Sscrofa11.1 | |
Downstream | ankyrin repeat and MYND domain containing 1 | ANKMY1 | 10.5kb | Anti-sense | Chr. 15 - 139485294 - 139531861 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
