Assay Details
Gene Symbol: MIR545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038609.1 | 1 | 42 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-374b | MIR374B | ssc-mir-374b | -51.5kb | Sense | Chr. X - 59551146 - 59551225 on NCBI Build Sscrofa11.1 |
Downstream | microRNA mir-374a | MIR374A | ssc-mir-374a | 87.0 | Sense | Chr. X - 59602875 - 59602954 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
