Assay Details
| Gene Symbol: LOC100626841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045015.1 | 2 - 3 | 230 | 107 | ||
| NR_045016.1 | 2 - 3 | 163 | 107 | |||
| GenBank mRNA | AK240197.1 | - | 2 - 3 | 163 | 107 | |
| AK347934.1 | - | 2 - 3 | 230 | 107 | ||
| AK348013.1 | - | 2 - 3 | 183 | 107 | ||
| AK352217.1 | - | 2 - 3 | 163 | 107 | ||
| AK398570.1 | - | 2 - 3 | 191 | 107 | ||
| AK398728.1 | - | 2 - 3 | 247 | 107 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adhesion molecule with Ig like domain 2 | AMIGO2 | -27.8kb | Anti-sense | Chr. 5 - 77642144 - 77645748 on NCBI Build Sscrofa11.1 | |
| Downstream | RNA polymerase II associated protein 3 | RPAP3 | 292.3kb | Anti-sense | Chr. 5 - 77968171 - 78066252 on NCBI Build Sscrofa11.1 |
More Information
Set Membership: |
|

LOC100626841
Genomic Map