Assay Details
| Gene Symbol: N4BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126990.5 | XP_003127038.2 | 2110 | 62 | ||
| GenBank mRNA | AK231450.1 | - | 2 - 3 | 975 | 62 | |
| AK389353.1 | - | 2 - 3 | 497 | 62 | ||
More Information
Set Membership: |
|

N4BP1
Genomic Map