Assay Details
Gene Symbol: RMDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005662992.3 | XP_005663049.1 | 453 | 64 | ||
XM_013996637.2 | XP_013852091.1 | 454 | 64 | |||
XM_013996638.2 | XP_013852092.1 | 350 | 64 | |||
XM_021089160.1 | XP_020944819.1 | 456 | 64 | |||
XM_021089161.1 | XP_020944820.1 | 455 | 64 | |||
GenBank mRNA | AK233488.1 | - | 3 - 4 | 349 | 64 | |
AK392414.1 | - | 3 - 4 | 438 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
