Assay Details
| Gene Symbol: SHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066254.1 | XP_020921913.1 | 1971 | 70 | ||
| GenBank mRNA | AK349535.1 | - | 4 - 5 | 592 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066254.1 | XP_020921913.1 | 1971 | 70 | ||
| GenBank mRNA | AK349535.1 | - | 4 - 5 | 592 | 70 | |
Set Membership: |
|