Assay Details
| Gene Symbol: PCBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358457.4 | XP_003358505.1 | 1174 | 56 | ||
| XM_005669618.3 | XP_005669675.1 | 1176 | 56 | |||
| XM_013981758.2 | XP_013837212.1 | 1048 | 56 | |||
| XM_013981759.2 | XP_013837213.1 | 1045 | 56 | |||
| XM_013981760.2 | XP_013837214.1 | 897 | 56 | |||
| XM_021068874.1 | XP_020924533.1 | 1168 | 56 | |||
| GenBank mRNA | AK235341.1 | - | 12 - 13 | 1100 | 56 | |
| AK395499.1 | - | 12 - 13 | 1107 | 56 | ||
More Information
Set Membership: |
|

PCBP4
Genomic Map