Assay Details
| Gene Symbol: PPM1J | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090756.1 | XP_020946415.1 | 1088 | 63 | ||
| XM_021090757.1 | XP_020946416.1 | 1061 | 63 | |||
| XM_021090758.1 | XP_020946417.1 | 989 | 63 | |||
| XM_021090759.1 | XP_020946418.1 | 878 | 63 | |||
| GenBank mRNA | AK394809.1 | - | 6 - 7 | 975 | 63 | |
More Information
Set Membership: |
|

PPM1J
Genomic Map