Assay Details
| Gene Symbol: WDR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081576.1 | XP_020937235.1 | 316 | 103 | ||
| XM_021081578.1 | XP_020937237.1 | 309 | 103 | |||
| XM_021081581.1 | XP_020937240.1 | 319 | 103 | |||
| GenBank mRNA | AK234626.1 | - | 283 | 103 | ||
More Information
Set Membership: |
|

WDR20
Genomic Map