Assay Details
| Gene Symbol: CNTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070955.1 | XP_020926614.1 | 602 | 86 | ||
| GenBank mRNA | AK231408.1 | - | 2 - 3 | 323 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CNTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070955.1 | XP_020926614.1 | 602 | 86 | ||
| GenBank mRNA | AK231408.1 | - | 2 - 3 | 323 | 86 | |
Set Membership: |
|