Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071309.1 | XP_020926968.1 | 2057 | 81 | ||
| GenBank mRNA | AK239668.1 | - | 10 - 11 | 2054 | 81 | |
More Information
Set Membership: |
|

TOMM70
Genomic Map