Assay Details
| Gene Symbol: ZKSCAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005661886.3 | XP_005661943.1 | 673 | 81 | ||
| XM_013995409.2 | XP_013850863.1 | 672 | 81 | |||
| XM_021086191.1 | XP_020941850.1 | 672 | 81 | |||
| GenBank mRNA | AK233041.1 | - | 2 - 3 | 637 | 81 | |
| AK390240.1 | - | 2 - 3 | 651 | 81 | ||
More Information
Set Membership: |
|

ZKSCAN1
Genomic Map