Assay Details
| Gene Symbol: CTCF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244660.1 | NP_001231589.1 | 5 - 6 | 1364 | 77 | |
| XM_021093368.1 | XP_020949027.1 | 9325 | 77 | |||
| XM_021093369.1 | XP_020949028.1 | 9420 | 77 | |||
| XM_021093370.1 | XP_020949029.1 | 9325 | 77 | |||
| XM_021093371.1 | XP_020949030.1 | 3499 | 77 | |||
| XM_021093372.1 | XP_020949031.1 | 9420 | 77 | |||
| XM_021093373.1 | XP_020949032.1 | 9325 | 77 | |||
| GenBank mRNA | AK234050.1 | - | 5 - 6 | 665 | 77 | |
| AK238742.1 | - | 2 - 3 | 182 | 77 | ||
| AK399712.1 | - | 7 - 8 | 1681 | 77 | ||
More Information
Set Membership: |
|

CTCF
Genomic Map