Assay Details
| Gene Symbol: HMGCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252215.1 | NP_001239144.1 | 2 - 3 | 120 | 63 | |
| XM_005657612.3 | XP_005657669.2 | 239 | 63 | |||
| XM_021076557.1 | XP_020932216.1 | 155 | 63 | |||
| GenBank mRNA | AK235559.1 | - | 2 - 3 | 120 | 63 | |
| AK400678.1 | - | 2 - 3 | 274 | 63 | ||
| GQ913656.1 | - | 2 - 3 | 108 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252215.1, XM_005657612.3 transcripts.
Set Membership: |
|

HMGCS1
Genomic Map