Assay Details
| Gene Symbol: DUSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256075.1 | NP_001243004.1 | 2 - 3 | 513 | 73 | |
| XM_021076556.1 | XP_020932215.1 | 851 | 73 | |||
| GenBank mRNA | AB490121.1 | - | 2 - 3 | 513 | 73 | |
| AK232967.1 | - | 2 - 3 | 752 | 73 | ||
| AK348088.1 | - | 2 - 3 | 782 | 73 | ||
| AK400206.1 | - | 1 - 2 | 118 | 73 | ||
More Information
Set Membership: |
|

DUSP1
Genomic Map