Assay Details
| Gene Symbol: CABYR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256771.3 | NP_001243700.2 | 2 - 3 | 209 | 116 | |
| NM_001348766.1 | NP_001335695.1 | 2 - 3 | 209 | 116 | ||
| NM_001348767.1 | NP_001335696.1 | 2 - 3 | 209 | 116 | ||
More Information
Set Membership: |
|

CABYR
Genomic Map