Assay Details
| Gene Symbol: AKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256779.1 | NP_001243708.1 | 6 - 7 | 641 | 66 | |
| XM_005655818.3 | XP_005655875.2 | 895 | 66 | |||
| XM_005655819.3 | XP_005655876.2 | 1015 | 66 | |||
| XM_005655820.3 | XP_005655877.2 | 911 | 66 | |||
| XM_013988560.2 | XP_013844014.2 | 879 | 66 | |||
| XM_013988561.2 | XP_013844015.2 | 809 | 66 | |||
| XM_013988563.2 | XP_013844017.2 | 929 | 66 | |||
| XM_021093202.1 | XP_020948861.1 | 809 | 66 | |||
| XM_021093204.1 | XP_020948863.1 | 645 | 66 | |||
| XM_021093205.1 | XP_020948864.1 | 903 | 66 | |||
| GenBank mRNA | HQ687754.1 | - | 6 - 7 | 641 | 66 | |
More Information
Set Membership: |
|

AKT2
Genomic Map