Assay Details
| Gene Symbol: GTSF1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258404.2 | NP_001245333.2 | 1 | 320 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001258404.2 transcripts.
Set Membership: |
|

GTSF1L
Genomic Map