Assay Details
| Gene Symbol: MMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044595.2 | NP_001038060.2 | 2 - 3 | 181 | 60 | |
| XM_021066259.1 | XP_020921918.1 | 365 | 60 | |||
| GenBank mRNA | AK233959.1 | - | 2 - 3 | 310 | 60 | |
| AK343305.1 | - | 2 - 3 | 312 | 60 | ||
| DQ656344.1 | - | 2 - 3 | 198 | 60 | ||
More Information
Set Membership: |
|

MMD
Genomic Map