Assay Details
| Gene Symbol: TRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260479.1 | NP_001247408.1 | 1 - 2 | 217 | 77 | |
| XM_005669803.3 | XP_005669860.1 | 472 | 77 | |||
| XM_021068420.1 | XP_020924079.1 | 441 | 77 | |||
| GenBank mRNA | JQ354941.1 | - | 1 - 2 | 217 | 77 | |
More Information
Set Membership: |
|

TRH
Genomic Map