Assay Details
| Gene Symbol: SEPT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278761.1 | NP_001265690.1 | 2 - 3 | 321 | 60 | |
| XM_021085714.1 | XP_020941373.1 | 2061 | 60 | |||
| GenBank mRNA | JX477173.1 | - | 2 - 3 | 321 | 60 | |
More Information
Set Membership: |
|

SEPT12
Genomic Map