Assay Details
| Gene Symbol: CDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285465.1 | NP_001272394.1 | 3 - 4 | 447 | 60 | |
| XM_003126260.5 | XP_003126308.1 | 514 | 60 | |||
| XM_005655594.2 | XP_005655651.1 | 554 | 60 | |||
| GenBank mRNA | AK231451.1 | - | 3 - 4 | 487 | 60 | |
| AK238749.1 | - | 3 - 4 | 489 | 60 | ||
| AK395994.1 | - | 3 - 4 | 541 | 60 | ||
| JX967576.1 | - | 3 - 4 | 450 | 60 | ||
More Information
Set Membership: |
|

CDK2
Genomic Map