Assay Details
| Gene Symbol: TSSK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287415.1 | NP_001274344.1 | 1 | 926 | 92 | |
| GenBank mRNA | KF640093.1 | - | 1 | 926 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TSSK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287415.1 | NP_001274344.1 | 1 | 926 | 92 | |
| GenBank mRNA | KF640093.1 | - | 1 | 926 | 92 | |
Set Membership: |
|