Assay Details
| Gene Symbol: ALDH3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315593.1 | NP_001302522.1 | 2 - 3 | 216 | 95 | |
| XM_021080123.1 | XP_020935782.1 | 95 | ||||
| XM_021080124.1 | XP_020935783.1 | 95 | ||||
| XM_021080126.1 | XP_020935785.1 | 95 | ||||
| XM_021080130.1 | XP_020935789.1 | 95 | ||||
| GenBank mRNA | AK240330.1 | - | 2 - 3 | 95 | ||
| KP735777.1 | - | 2 - 3 | 216 | 95 | ||
| KP735784.1 | - | 2 - 3 | 216 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021080130.1 transcripts.
Set Membership: |
|

ALDH3B1
Genomic Map